Learn More
Specifications
Specifications
| Antigen | CD3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | UCHT1 |
| Conjugate | PerCP |
| Description | Mouse monoclonal antibody raised against native CD3. |
| Dilution | Flow Cytometry (10 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide) |
| Gene Alias | CD3-DELTA/T3D |
| Gene Symbols | CD3D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.