Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300A, Mouse, Clone: MEM-260, Abnova™

Catalog No. 89111620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-620 Supplier Abnova Corporation Supplier No. MAB3834
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD300A.

The CMRF35 antigen (CMRF35A; MIM 606786), which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. CMRF35H is recognized by the same antibody and is distinct from CMRF35 (Green et al., 1998 [PubMed 9701027]).[supplied by OMIM

Specifications

Antigen CD300A
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-260
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD300A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CD300A
Gene Alias CMRF-35-H9/CMRF-35H/CMRF35H/CMRF35H9/IGSF12/IRC1/IRC2/IRp60
Gene Symbols CD300A
Host Species Mouse
Immunogen Native purified CD300A from human HPB acute lymphoid leukaemia cell line.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11314
Test Specificity This antibody reacts with CD300a, a 60 KDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.