Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD320, Mouse, Clone: 4F2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CD320.
Supplier: Abnova Corporation H00051293M04A
Description
Sequence: MSGGWMAQVGAWRTGALGLALLLLLGLGLGLEAAASPLSTPTSAQAAGPSSGSCPPTKFQCRTSGLCVPLTWRCDRDLDCSDGSDEEECRIEPCTQKGQCPPPPGLPCPCTGVSDCSGGTDKKLRNCSRLACLAGELRCTLSDDCIPLTWRCDGHPDCPDSSDELGCGTNEILPEGDATTMGPPVTLESVTSLRNATTMGPPVTLESVPSVGNATSSSAGDQSGSPTAYGVIAAAAVLSASLVTATLLLLSWLRAQERLRPLGLLVAMKESLLLSEQKTSLPSpecifications
CD320 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC000668 | |
CD320 | |
CD320 (AAH00668,1 a.a. ∽ 282 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
51293 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
4F2 | |
Mouse monoclonal antibody raised against a full-length recombinant CD320. | |
CD320 | |
8D6/8D6A | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction