Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALB, Mouse, Clone: 4H5E7, Abnova™

Catalog No. 89113420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-420 Supplier Abnova Corporation Supplier No. MAB8296
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant CD34.

CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110kDa that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM

Specifications

Antigen ALB
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4H5E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CD34.
Dilution Western Blot (1:200-1:1000) Immunofluorescence (1:25-1:100) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene CD34
Gene Symbols CD34
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human CD34.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.