Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34, Mouse, FITC, Clone: 4H11[APG], Abnova™

Catalog No. 89111809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-809 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-809 Supplier Abnova Corporation Supplier No. MAB4412
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD34.

CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110kDa that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM

Specifications

Antigen CD34
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4H11[APG]
Conjugate FITC
Description Mouse monoclonal antibody raised against native CD34.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD34
Gene Symbols CD34
Host Species Mouse
Immunogen Native purified CD34 from permanent human cell line derived from peripheral leucocytes of a patient suffering from chronic myeloid leukaemia.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Test Specificity This antibody reacts with Class III epitope on CD34 (Mucosialin), a 110-115 KDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.