Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34, Mouse, PE, Clone: ICO115, Abnova™

Catalog No. 89109442 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-442 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-442 Supplier Abnova Corporation Supplier No. MAB11277
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD34.

CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110kDa that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone ICO115
Conjugate PE
Description Mouse monoclonal antibody raised against CD34.
Dilution Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene CD34
Gene Symbols CD34
Host Species Mouse
Immunogen Blast cells from a chronic myeloid leukemia patient
Purification Method Protein G affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.