Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Mouse anti-Human, Alexa Fluor 647, Clone: E11, BD

Catalog No. BDB565329 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565329 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565329 Supplier BD Biosciences Supplier No. 565329

Mouse Monoclonal Antibody

The E11 monoclonal antibody specifically binds to CD35. CD35 is also known as Complement receptor type 1 (CR1), C3b/C4b receptor, C3BR, C4BR, Immune adherence receptor, or KN. CD35 is a type I transmembrane glycoprotein that exists in four allelic forms of 160, 190, 220 and 250 kDa. CD35 serves as a receptor for complement fragments C3b, iC3b, C3dg, C4b, iC3, and iC4. It enhances phagocytosis by neutrophils and monocytes and regulates complement activation. It is expressed on erythrocytes, granulocytes, monocytes, B cells, and some dendritic cells, T cells, and NK cells. It binds complement components C3b and C4b, mediating. This antibody cannot inhibit the phagocytic capacity of granulocytes. The CD35 antibody is useful in studies of cells that express complement receptors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD35
Applications Flow Cytometry
Classification Monoclonal
Clone E11
Conjugate Alexa Fluor 647
Description CR1; Complement receptor type 1; C3b/C4b receptor; C3BR; C4BR; KN
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cells of the Monocyte Lineage
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.