Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd36, Mouse, Clone: D-2712, Abnova™

Catalog No. 89110932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-932 Supplier Abnova Corporation Supplier No. MAB2325
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Cd36.

Specifications

Antigen Cd36
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D-2712
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Cd36.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Cd36
Gene Alias FAT/GPIV/Scarb3
Gene Symbols Cd36
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Cd36.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12491
Test Specificity This antibody is specific to CD36.
Target Species Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgA κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.