Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD36/SR-B3, Clone: 1E8., Novus Biologicals™
SDP

Catalog No. NBP24525102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24525102 0.2 mg
NBP24525101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP24525102 Supplier Novus Biologicals Supplier No. NBP2452510.2MG

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

CD36/SR-B3 Monoclonal antibody specifically detects CD36/SR-B3 in Human samples. It is validated for Flow Cytometry, SDS-Page, Immunofluorescence.

Specifications

Antigen CD36/SR-B3
Applications Flow Cytometry, SDS-Page, Immunofluorescence
Classification Monoclonal
Clone 1E8.
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias CD36 antigen, CD36 molecule (thrombospondin receptor), FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor
Gene Symbols CD36
Host Species Mouse
Immunogen Human CD36 from platelets
Molecular Weight of Antigen 85 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 948
Test Specificity Recognizes a protein of 80kDa-90kDa, identified as CD36. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischemia observed in sickle cell disease and cerebral malaria. Note that 1-4% of Japanese and East Asia population lack CD36.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.