Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD369 Mouse anti-Human, Alexa Fluor 647, Clone: 15E2, BD

Catalog No. BDB564855 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564855 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564855 Supplier BD Biosciences Supplier No. 564855
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 15E2 monoclonal antibody specifically binds to CD369 which is also known as CLEC7A (C-type lectin domain family 7 member A), Dectin-1 (Dendritic cell-associated C-type lectin 1), or BGR (Beta-glucan receptor). CD369 is a type II transmembrane C-type lectin that contains a C-terminal stalk with a carbohydrate recognition domain (CRD) in the extracellular domain, a transmembrane segment, and an ITAM containing cytoplasmic tail. This protein is highly expressed on dendritic cells. It is also expressed on B cells, monocytes, eosinophils, mast cells, macrophages, endothelial cells, and is lowly expressed on some subsets of T cells. CD369 binds to beta-glucan polymers and functions as a pattern recognition receptor in innate immune response to fungal and bacterial pathogens. The 15E2 antibody can reportedly stimulate dendritic cells to costimulate immune responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD369
Applications Flow Cytometry
Classification Monoclonal
Clone 1.50E+03
Conjugate Alexa Fluor 647
Description CLEC7A; CLC7A; BGR; CLECSF12; CANDF4
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Dectin-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.