Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CD37, Clone: MB-1,

Catalog No. MABF913 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF913 Supplier MilliporeSigma™ Supplier No. MABF913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD37 clone: MB-1 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Leukocyte antigen CD37 (UniProt P11049; also known as CD37, Tetraspanin-26, Tspan-26) is encoded by the CD37 (also known as TSPAN26) gene (Gene ID 951) in human. CD37 is a 4-transmembrane (a.a. 18-38, 60-74, 86-111, 242-266) tetraspanin superfamily member with both its N- and C-terminal ends exposed cytoplasmically (a.a. 1-17, 267-281). While most tetraspanins are ubiquitously expressed, CD37 is predominently expressed on antigen-presenting cells, with highest expression on B lymphocytes. CD37 is essential for B cell survival and Cd37-/- mice showed greatly reduced IgG-secreting plasma cells in lymphoid organs due to increased apoptosis in the spleen germinal centers, where B cells proliferate. On the other hand, CD37 is also found to play a negative role against T cell proliferation, macrophages interleukin-6 production, and dendritic cells (DCs) antigen presentation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD37
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MB-1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P11049
Gene Symbols CD37; TSPAN26
Host Species Mouse
Immunogen Lymph node cells from a lymphoma patient.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001765.1
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.