Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38, Mouse, Clone: AT1, Abnova™
Mouse monoclonal antibody raised against native CD38.
Supplier: Abnova Corporation MAB6909
Description
CD38 is a novel multifunctional ectoenzyme widely expressed in cells and tissues especially in leukocytes. CD38 also functions in cell adhesion, signal transduction and calcium signaling. [provided by RefSeq
Specifications
CD38 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CD38 | |
CD38 | |
Native purified CD 38 from human T cell tumor cells: CEM. | |
RUO | |
952 | |
Store at 4°C. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
AT1 | |
Mouse monoclonal antibody raised against native CD38. | |
In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate) | |
T10 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction