Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3E, Mouse, Clone: CA-3, Abnova™

Catalog No. 89110428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-428 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-428 Supplier Abnova Corporation Supplier No. MAB1433
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD3E.

The protein encoded by this gene is the CD3-epsilon polypeptide, which together with CD3-gamma, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. Defects in this gene cause immunodeficiency. This gene has also been linked to a susceptibility to type I diabetes in women. [provided by RefSeq

Specifications

Antigen CD3E
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CA-3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD3E.
Dilution Immunohistochemistry (Frozen sections) (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene CD3E
Gene Alias FLJ18683/T3E/TCRE
Gene Symbols CD3E
Host Species Mouse
Immunogen Native purified CD3E from human thymocytes followed by Sezary T cells.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Human, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.