Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Mouse, Clone: 10C12, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CD4.
Supplier: Abnova Corporation MAB8338
Description
Sequence: LSEKKTCQCPHRFQKTCSPISpecifications
CD4 | |
Monoclonal | |
Unconjugated | |
ELISA (10-20 ug/mL) Immunofluorescence (5-10 ug/mL) Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
CD4 | |
CD4 | |
A synthetic peptide corresponding to human CD4. | |
RUO | |
920 | |
Human | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
10C12 | |
Mouse monoclonal antibody raised against synthetic peptide of CD4. | |
In PBS, pH 7.4 | |
CD4mut | |
Mouse | |
100 μg | |
Primary | |
This antibody recognizes an epitope within amino acids 414-433 of the cytoplasmic tail region of human CD4. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction