Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse, Clone: 15-8A2, Abnova™

Catalog No. 89112212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-212 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-212 Supplier Abnova Corporation Supplier No. MAB5300
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD4.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 15-8A2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD4.
Dilution Flow Cytometry (1:10) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.01% sodium azide)
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen Native purified from WAG/Rij spleen cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920/12504
Test Specificity The monoclonal antibody recognizes rat CD4 antigen (MW 52KDa) and is reactive with all common inbred strains.
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.