Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse, Clone: 8A5, Abnova™

Catalog No. 89113440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-440 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-440 Supplier Abnova Corporation Supplier No. MAB8337
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CD4.

Sequence: RFLPADQMNVADPQPLASCS

Specifications

Antigen CD4
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 8A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CD4.
Dilution ELISA (10-20 ug/mL) Immunofluorescence (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene CD4
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen A synthetic peptide corresponding to human CD4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity This antibody recognizes the post 3'-UGA codon region of human CD4.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.