Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Mouse, Clone: CA-4, Abnova™
Mouse monoclonal antibody raised against recombinant CD4.
Supplier: Abnova Corporation MAB1434
Specifications
CD4 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Frozen sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
CD4 | |
CD4 | |
Recombinant protein corresponding to human T-cell CEM cells of human CD4. | |
100 μg | |
Primary | |
Human, Rat | |
Lyophilized |
Immunohistochemistry (Frozen) | |
CA-4 | |
Mouse monoclonal antibody raised against recombinant CD4. | |
Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
CD4mut | |
Mouse | |
Affinity purification | |
RUO | |
920 | |
Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction