Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse, Clone: HP2/6, Abnova™

Catalog No. 89112012 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-012 Supplier Abnova Corporation Supplier No. MAB4960
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD4.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone HP2/6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD4.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide)
Gene CD4
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen Japanese white rabbit thymocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.