Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Mouse, Clone: MEM-241, Abnova™
Mouse monoclonal antibody raised against partial recombinant CD4.
Supplier: Abnova Corporation MAB3837
Specifications
CD4 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (1 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
CD4 | |
CD4 | |
Recombinant Fc fusion protein corresponding to 2 N-terminus domains of human CD4. | |
RUO | |
920 | |
Human | |
Liquid |
Flow Cytometry, Western Blot | |
MEM-241 | |
Mouse monoclonal antibody raised against partial recombinant CD4. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CD4mut | |
Mouse | |
100 μg | |
Primary | |
This antibody recognizes CD4 antigen, a 55 KDa transmebrane glycoprotein expressed on a subset of T lymphocytes (helper T-cells) and also on monocytes, tissue macrophages and granulocytes. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction