Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CD4, Clone: RPA-T4, PE,

Catalog No. MABF1720 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1720 500 Tests
MABF1718 25 Tests
MABF1719 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MABF1720 Supplier MilliporeSigma™ Supplier No. MABF1720
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD4 clone: RPA-T4 in Human samples, and it is validated for Flow Cytometry

The RPA-T4 antibody reacts with human CD4, a 59kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The RPA-T4 antibody is widely used as a phenotypic marker for human CD4 expression, and is cross-reactive with Chimpanzee CD4. This antibody recognizes a different epitope, and thus does not block binding of, the alternative Anti-Human CD4 antibody clone OKT4 (Reinherz EL, et al. 1979. Proc. Natl. Acad. Sci. 76:4061-4065).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone RPA-T4
Conjugate PE
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P01730
Gene Symbols CD4
Host Species Mouse
Immunogen Immunogen information for this antibody is proprietary information.
Purification Method Purified by size-exclusion chromatography
Quantity 500 Tests
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.