Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse, FITC, Clone: MEM-241, Abnova™

Catalog No. 89111863 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-863 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-863 Supplier Abnova Corporation Supplier No. MAB4510
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CD4.

Specifications

Antigen CD4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-241
Conjugate FITC
Description Mouse monoclonal antibody raised against partial recombinant CD4.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD4
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to partial human CD4.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity This antibody recognizes CD4 antigen, a 55 KDa transmebrane glycoprotein expressed on a subset of T lymphocytes (helper T-cells) and also on monocytes, tissue macrophages and granulocytes.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.