Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Mouse, PE, Clone: RFT-4g, Abnova™
Mouse monoclonal antibody raised against native CD4.
Supplier: Abnova Corporation MAB6013
Specifications
CD4 | |
Monoclonal | |
PE | |
Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
CD4 | |
CD4 | |
Native purified CD4 from human thymocytes and E-rosetted lymphocytes. | |
RUO | |
920 | |
Human | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
RFT-4g | |
Mouse monoclonal antibody raised against native CD4. | |
In PBS (0.09% sodium azide) | |
CD4mut | |
Mouse | |
100 Reactions | |
Primary | |
Specificity human CD4 | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction