Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse, PE-Cy5.5, Clone: RFT-4g, Abnova™

Catalog No. 89112629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-629 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-629 Supplier Abnova Corporation Supplier No. MAB6018
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD4.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone RFT-4g
Conjugate PE-Cyanine5.5
Description Mouse monoclonal antibody raised against native CD4.
Dilution Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CD4
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen Native purified CD4 from human thymocytes and E-rosetted lymphocytes.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity Specificity human CD4
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.