Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41/CD61, Mouse, Clone: IVA30, Abnova™

Catalog No. 89108309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-309 Supplier Abnova Corporation Supplier No. MAB0987
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD41/CD61.

Specifications

Antigen CD41/CD61
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone IVA30
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD41/CD61.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias INTB3
Gene Symbols ITGB3
Host Species Mouse
Immunogen Native purified CD41/CD61 from bovine trombocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282642/515011
Test Specificity This antibody reacts with CD41/CD61 complex (bovine), one of the earliest markers of the megakaryocytic lineage.
Target Species Bovine
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.