Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42a Mouse anti-Human, Brilliant Violet 421, Clone: ALMA.16, BD

Catalog No. BDB565444 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565444 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565444 Supplier BD Biosciences Supplier No. 565444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 17-22kD, single chain integral membrane glycoprotein, also known as GPIX. CD42a forms a noncovalently linked complex (GPIb/GPIX/GPV) with CD42b, CD42c and CD42d. It is expressed on platelets and megakaryocytes and is absent on the platelets of patients with Bernadr-Soulier Syndrome (BSS). Although the CD42a function is not fully understood, GPIX glycoprotein is important for the assembly and membrane expression of the complex and for the maintenance of the functional conformation of CD42b (GPIb).

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications Flow Cytometry
Classification Monoclonal
Clone ALMA.16
Conjugate Brilliant Violet 421
Description GPIX; GP9; Glycoprotein IX
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.