Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44, Mouse, Clone: DF 1485, Abnova™

Catalog No. 89112218 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-218 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-218 Supplier Abnova Corporation Supplier No. MAB5315
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD44.

The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). This protein participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis. [provided by RefSeq

Specifications

Antigen CD44
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone DF 1485
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD44.
Dilution Immunohistochemistry (1:5-1:10) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant
Gene CD44
Gene Alias CDW44/CSPG8/ECMR-III/HCELL/IN/LHR/MC56/MDU2/MDU3/MGC10468/MIC4/MUTCH-I/Pgp1
Gene Symbols CD44
Host Species Mouse
Immunogen Native purified human CD44.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Test Specificity HCAM, CD44 is a transmembrane glycoprotein which is expressed by lymphocytes and several normal epithelial cells. The CD44 molecule is involved in lymphocyte homing, T-lymphocyte actvitation, interaction with hyaluronic acid and may act as an adhesion molecule. Standard CD 44 (CD44s) is composed of the products of exons 1-5 a less favourable prognosis in gastrointestinal cancers, the loss of CD44s expression predicts unfavourable outcome in bladder cancers, squamous cell carcinomas of the skin, prostatic adenocarcinomas and neuroblastomas. In immunoblotting this antibody reacts with a glycoprotein isolated from hemopoietic cells and epithelials cells with a respective molecular weight of 29-37 and 51 kD.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.