Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CD44, Clone: HP2/9, Abnova™

Catalog No. 89112031 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-031 Supplier Abnova Corporation Supplier No. MAB4981
Add to Cart
Add to Cart

Specifications

Antigen CD44
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone HI44a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD44. The monoclonal antibody is directed against the CD44- antigen, which is mainly expressed on leucocytes and erythrocytes. Very few types of tissue or cells lack CD44. Cells and tissues that lack CD44 include platelets, hepatocytes, certain lymphoid cell lines, cardiac muscle, kidney tubular epithelium, testis and portions of the skin. The CD44 antigen is present on approximately 90% of lymphocytes, monocytes, and granulocytes and in lower amounts on thymocytes, fibroblasts, and erythrocytes.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide)
Gene CD44
Gene Alias CDW44/CSPG8/ECMR-III/HCELL/IN/LHR/MC56/MDU2/MDU3/MGC10468/MIC4/MUTCH-I/Pgp1
Gene Symbols CD44
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.