Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44, Mouse, FITC, Clone: MEM-263, Abnova™

Catalog No. 89136794 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-794 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-794 Supplier Abnova Corporation Supplier No. MAB9988
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD44.

Specifications

Antigen CD44
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-263
Conjugate FITC
Description Mouse monoclonal antibody raised against CD44.
Dilution Flow cytometry (20 ul reagent / 100 ul of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide, 0.2% (w/v) BSA)
Host Species Mouse
Immunogen COS-7 cells (African Green Monkey)
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody reacts with extracellular (N-terminal) domain of standard CD44, a 80-95 kDa transmembrane glycoprotein (hyaladherin family)present on the most of cells and tissues (leukocytes, endothelial cells, mesenchymal cells,etc.); it is negative on platelets and hepatocytes.
Target Species Canine, Human, Monkey, Pig
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.