Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA (PTPRC), Mouse, Clone: 158-4D3, Abnova™

Catalog No. 89109462 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-462 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-462 Supplier Abnova Corporation Supplier No. MAB11299
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD45RA (PTPRC).

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus belongs to receptor type PTP. This gene is specifically expressed in hematopoietic cells. This PTP has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. It functions through either direct interaction with components of the antigen receptor complexes, or by activating various Src family kinases required for the antigen receptor signaling. This PTP also suppresses JAK kinases, and thus functions as a regulator of cytokine receptor signaling. Four alternatively spliced transcripts variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq

Specifications

Antigen CD45RA (PTPRC)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Monoclonal
Clone 158-4D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CD45RA (PTPRC).
Dilution Flow Cytometry (1 ug/106 cells) Immunocytochemistry/Immunofluorescence (1:10-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500) Immunoprecipitation (1:10-1:500) The optimal working dilution should b
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene PTPRC
Gene Alias B220/CD45/CD45R/GP180/LCA/LY5/T200
Gene Symbols PTPRC
Host Species Mouse
Immunogen Stimulated human leukocytes.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.