Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47, Mouse, Clone: BRIC126, Abnova™

Catalog No. 89112202 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-202 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-202 Supplier Abnova Corporation Supplier No. MAB5279
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native purified CD47.

This gene encodes a membrane protein, which is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. The encoded protein is also a receptor for the C-terminal cell binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. This gene has broad tissue distribution, and is reduced in expression on Rh erythrocytes. Four alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CD47
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone BRIC126
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native purified CD47.
Dilution Flow Cytometry (10 ul of the suggested working dilution to label 106 cells in 100 ul) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycerine, pH 7.8 (0.09% sodium azide)
Gene CD47
Gene Alias IAP/MER6/OA3
Gene Symbols CD47
Host Species Mouse
Immunogen Native purified CD47 from human erythrocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 961
Test Specificity This antibody recognizes the human CD47 cell surface glycoprotein, a heavily N-glycosylated 47-52kD molecule. CD47 is expressed on all cells and tissues so far examined, although expression is reduced on erythrocytes of the rare Rh null phenotype.
Target Species Bovine, Canine, Human, Pig
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.