Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48, Mouse, APC, Clone: MEM-102, Abnova™

Catalog No. 89111946 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-946 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-946 Supplier Abnova Corporation Supplier No. MAB4635
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD48.

BLAST1 is the designation used for an activation-associated cell surface glycoprotein of 40 to 45kDa expressed primarily in mitogen-stimulated human lymphocytes. The protein sequence predicted by the cDNA encoding BLAST1 indicates that BLAST1 is a member of the immunoglobulin supergene family. Yokoyama (1991) identified the BLAST1 activation/adhesion molecule as CD48.[supplied by OMIM

Specifications

Antigen CD48
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-102
Conjugate APC
Description Mouse monoclonal antibody raised against native CD48.
Dilution Flow Cytometry (10 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD48
Gene Alias BCM1/BLAST/BLAST1/MEM-102/SLAMF2/hCD48/mCD48
Gene Symbols CD48
Host Species Mouse
Immunogen Native purified CD48 from Raji Burkitt's lymphoma cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 962
Test Specificity This antibody reacts with CD48 antigen (Blast-1), a 40-47 KDa GPI-anchored membrane protein (immunoglobulin supergene family) widely expressed on hematopoietic cells; it is negative on granulocytes, platelets and erythrocytes.
Target Species Human, Non-human Primate
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.