Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d VLA, Mouse, Clone: HP2.1, Abnova™

Catalog No. 89135749 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-749 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-749 Supplier Abnova Corporation Supplier No. MAB4729
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD49d VLA.

Specifications

Antigen CD49d VLA
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone HP2.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD49d VLA.
Dilution Immunofluorescence (2ug/5x105 cells/test) Flow Cytometry (2ug/5x105 cells/test) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1mg/mL BSA)
Host Species Mouse
Immunogen Native purified from JM leukemia line.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The molecular weight of the recognized antigen is 150KDa. CD49d antigen is the integrin alpha-4 chain non covalently associated with the integrin beta-1-chain (CD29 molecule) forming the VLA4 complex. Reacts with monocytes, T cells, B cells, thymocytes and Langerhans cells.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.