Learn More
CD5 Mouse anti-Human, APC-R700, Clone: UCHT2, BD
Mouse Monoclonal Antibody
$178.00 - $378.00
Specifications
Antigen | CD5 |
---|---|
Clone | UCHT2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-R700 |
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).
Specifications
CD5 | |
Flow Cytometry | |
APC-R700 | |
RUO | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CD5 antigen (p56-62); T1; Tp67; LEU1; Lymphocyte antigen T1/Leu-1 |
UCHT2 | |
Monoclonal | |
Mouse | |
Human | |
Human T cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.