Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5, Mouse, Clone: C5/473, Abnova™

Catalog No. 89109414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-414 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-414 Supplier Abnova Corporation Supplier No. MAB11244
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CD5.

Specifications

Antigen CD5
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone C5/473
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CD5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Bioreactor concentrate (0.05% sodium azide)
Gene CD5
Gene Alias LEU1/T1
Gene Symbols CD5
Host Species Mouse
Immunogen Recombinant protein corresponding to human CD5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 921
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.