Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52, Mouse, Clone: HI186, Abnova™

Catalog No. 89108272 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-272 Supplier Abnova Corporation Supplier No. MAB0944
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD52.

Specifications

Antigen CD52
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone HI186
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD52.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CD52
Gene Alias CDW52
Gene Symbols CD52
Host Species Mouse
Immunogen Native purified CD52 from human tonsil.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1043
Test Specificity This antibody reacts with CD52 (CAMPATH-1), a 21-28 KDa glycoprotein containing a large N-linked carbohydrate moiety; mature CD52 molecule is actually much smaller (approx. 8-9 KDa). CD52 is expressed at high levels on lymphocytes, monocytes/macrophages and in male reproductive tract.
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.