Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55, Mouse, Clone: 67, Abnova™

Catalog No. 89112173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-173 Supplier Abnova Corporation Supplier No. MAB5221
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD55.

This gene encodes a protein involved in the regulation of the complement cascade. The encoded glycoprotein is also known as the decay-accelerating factor (DAF); binding of DAF to complement proteins accelerates their decay, disrupting the cascade and preventing damage to host cells. Antigens present on the DAF glycoprotein constitute the Cromer blood group system (CROM). Two alternatively spliced transcripts encoding different proteins have been identified. The predominant transcript encodes a membrane-bound protein expressed on cells exposed to plasma component proteins but an alternatively spliced transcript produces a soluble protein present at much lower levels. Additional, alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Specifications

Antigen CD55
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 67
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD55.
Dilution Flow Cytometry (1:10-1:25) Immunohistochemistry (Frozen sections) (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CD55
Gene Alias CR/CROM/DAF/TC
Gene Symbols CD55
Host Species Mouse
Immunogen Native purified CD55 from K562 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1604
Test Specificity This antibody recognizes the human CD55 cell surface antigen, a GPI linked molecule also known as decay accelerating factor (DAF). CD55 is expressed by a wide range of cell types. Erythrocyte expression is very weak.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.