Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6, Mouse, Clone: C6/372, Abnova™

Catalog No. 89109498 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-498 Supplier Abnova Corporation Supplier No. MAB11335
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD6.

CD6 is a monomeric 105- or 130kDa membrane glycoprotein that is involved in T-cell activation. The size difference between the 2 CD6 forms is due to differences in phosphorylation (Robinson et al., 1995 [PubMed 7589069]).[supplied by OMIM

Specifications

Antigen CD6
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C6/372
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CD6.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1:100-1:200) Immunohistochemistry (1:100-1:200; 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (1:250-1:500) The optimal working dilution should be determin
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene CD6
Gene Alias FLJ44171/TP120
Gene Symbols CD6
Host Species Mouse
Immunogen Human rheumatoid synovial T cell line ST-1.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 923
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.