Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6, Mouse, Clone: MEM-98, Abnova™

Catalog No. 89111627 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-627 Supplier Abnova Corporation Supplier No. MAB3841
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD6.

CD6 is a monomeric 105- or 130kDa membrane glycoprotein that is involved in T-cell activation. The size difference between the 2 CD6 forms is due to differences in phosphorylation (Robinson et al., 1995 [PubMed 7589069]).[supplied by OMIM

Specifications

Antigen CD6
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-98
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD6.
Dilution Flow Cytometry (2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CD6
Gene Alias FLJ44171/TP120
Gene Symbols CD6
Host Species Mouse
Immunogen Native purified human CD6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 923
Test Specificity This antibody reacts with CD6, a 100-130 kD single chain transmembrane glycoprotein expressed on T and B lymphocytes subsets, thymocytes, and acute lymphocytic leukemia cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.