Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L, Mouse, Clone: IVA94, Abnova™

Catalog No. 89108310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-310 Supplier Abnova Corporation Supplier No. MAB0988
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD62L.

Specifications

Antigen CD62L
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone IVA94
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD62L.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SELL
Gene Symbols SELL
Host Species Mouse
Immunogen Native purified CD62L from bovine leukocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281485
Test Specificity This antibody reacts with CD62L antigen (bovine).
Target Species Bovine
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.