Learn More
IgM Mouse anti-Human, Unlabeled, Clone: 145-8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 345160
Description
Anti-IgM, clone 145-8, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with lymph node cells from BALB/c mice immunized with the human myeloma μ chain.
The Anti-IgM antibody is specific for all subclasses of human IgM heavy chains.

Specifications
IgM | |
Monoclonal | |
50μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide | |
Human myeloma μ chain | |
RUO | |
3507 | |
Store at 2-8°C. | |
IgG1 κ |
Flow Cytometry | |
145-8 | |
Unconjugated | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.