Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63, Mouse, PerCP, Clone: MEM-259, Abnova™

Catalog No. 89112901 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-901 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-901 Supplier Abnova Corporation Supplier No. MAB6493
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD63.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins. It may function as a blood platelet activation marker. Deficiency of this protein is associated with Hermansky-Pudlak syndrome. Also this gene has been associated with tumor progression. The use of alternate polyadenylation sites has been found for this gene. Alternative splicing results in multiple transcript variants encoding different proteins. [provided by RefSeq

Specifications

Antigen CD63
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MEM-259
Conjugate PerCP
Description Mouse monoclonal antibody raised against native CD63.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD63
Gene Alias LAMP-3/ME491/MLA1/OMA81H/TSPAN30
Gene Symbols CD63
Host Species Mouse
Immunogen Native purified CD63 from HPB-ALL T cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 967
Test Specificity This antibody reacts with CD63 (LAMP-3), a 40-60 KDa tetraspan glycoproteinexpressed by granulocytes, platelets, T cells, monocytes/macrophages and endothelialcells. Cell surface exposition of CD63 is usually activation-dependent.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.