Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69, Mouse, Clone: FN50, Abnova™

Catalog No. 89112879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-879 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-879 Supplier Abnova Corporation Supplier No. MAB6461
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD69.

This gene encodes a member of the calcium dependent lectin superfamily of type II transmembrane receptors. Expression of the encoded protein is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants

Specifications

Antigen CD69
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone FN50
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD69.
Dilution Flow Cytometry (5 ug/mL) Functional Study Immunohistochemistry The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CD69
Gene Alias CLEC2C
Gene Symbols CD69
Host Species Mouse
Immunogen Native purified CD69 from human B lymphocytes.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 969
Test Specificity This antibody recognizes CD69, an lymphocyte early activation marker.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.