Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7, Mouse, APC, Clone: MEM-186, Abnova™

Catalog No. 89111879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-879 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-879 Supplier Abnova Corporation Supplier No. MAB4535
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD7.

This gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily. This protein is found on thymocytes and mature T cells. It plays an essential role in T-cell interactions and also in T-cell/B-cell interaction during early lymphoid development. [provided by RefSeq

Specifications

Antigen CD7
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-186
Conjugate APC
Description Mouse monoclonal antibody raised against native CD7.
Dilution Flow Cytometry (10 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD7
Gene Alias GP40/LEU-9/TP41/Tp40
Gene Symbols CD7
Host Species Mouse
Immunogen Native purified CD7 from human acute myelogenous leukaemia cell line KG-1.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 924
Test Specificity The antibody reacts with CD7, a 40 KDa type I transmembrane glycoprotein expressed on peripheral blood T lymphocytes, NK-cells, hematopoietic progenitors, monocytes (weakly) and also on acute lymphocytic leukemia.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.