Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7, Mouse, Clone: LP15, Abnova™

Catalog No. 89113714 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-714 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-714 Supplier Abnova Corporation Supplier No. MAB9646

Mouse monoclonal antibody raised against partial recombinant CD7.

This gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily. This protein is found on thymocytes and mature T cells. It plays an essential role in T-cell interactions and also in T-cell/B-cell interaction during early lymphoid development. [provided by RefSeq

Specifications

Antigen CD7
Applications Immunohistochemistry
Classification Monoclonal
Clone LP15
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CD7.
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (1% fetal calf serum, 0.09% sodium azide)
Gene CD7
Gene Alias GP40/LEU-9/TP41/Tp40
Gene Symbols CD7
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of human CD7.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 924
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.