Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd72.1, Mouse, Clone: 10-1.D2, Abnova™

Catalog No. 89111285 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-285 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-285 Supplier Abnova Corporation Supplier No. MAB3237
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cd72.1.

Specifications

Antigen Cd72.1
Applications Flow Cytometry
Classification Monoclonal
Clone 10-1.D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cd72.1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Cd72
Gene Alias CD72c/Ly-19/Ly-32/Ly-m19/Lyb-2
Gene Symbols Cd72
Host Species Mouse
Immunogen DBA/2 mouse spleen cells.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12517
Test Specificity Mouse CD72.1/Lyb-2.1, 45 KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.