Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD72, Mouse, PE, Clone: 3F3, Abnova™

Catalog No. 89111838 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-838 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-838 Supplier Abnova Corporation Supplier No. MAB4463
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD72.

Specifications

Antigen CD72
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 3F3
Conjugate PE
Description Mouse monoclonal antibody raised against native CD72.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD72
Gene Alias CD72b/LYB2
Gene Symbols CD72
Host Species Mouse
Immunogen Native purified CD72 from normal human lymphocytes from lymph node.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 971
Test Specificity This antibody 3F3 reacts with CD72, a 39-43 KDa type II membrane glycoprotein (C-type lectin family). CD72 is a pan-B cell marker expressed throughout the B lymphocytes diferentiation with the exception of plasma cells; it is also present on follicular dendritic cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.