Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74, Mouse, Clone: PIN.1, Abnova™

Catalog No. 89112957 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-957 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-957 Supplier Abnova Corporation Supplier No. MAB6623
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CD74.

Specifications

Antigen CD74
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PIN.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CD74.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide)
Gene CD74
Gene Alias DHLAG/HLADG/Ia-GAMMA
Gene Symbols CD74
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to invariant chain of human CD74.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 972
Test Specificity Detects∼33-35KDa protein doublet corresponding to the molecular mass of the p33 and p35 forms of human CD74 on SDS PAGE immunoblots.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.