Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79A, Mouse, APC, Clone: HM57, Abnova™

Catalog No. 89111867 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-867 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-867 Supplier Abnova Corporation Supplier No. MAB4515
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CD79A.

The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component, surface immunoglobulin (Ig). Surface Ig noncovalently associates with two other proteins, Ig-alpha and Ig-beta, which are necessary for expression and function of the B-cell antigen receptor. This gene encodes the Ig-alpha protein of the B-cell antigen component. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Specifications

Antigen CD79A
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone HM57
Conjugate APC
Description Mouse monoclonal antibody raised against synthetic peptide of CD79A.
Dilution Flow Cytometry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD79A
Gene Alias IGA/MB-1
Gene Symbols CD79A
Host Species Mouse
Immunogen A synthetic peptide corresponding to human CD79A.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 973
Test Specificity This antibody interacts with CD79a (Ig alpha), a 40-45 KDa subunit of B cell antigen-specific receptor (BCR) and its early developmental forms.
Target Species Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Opossum, Pig, Rabbit, Rat
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.