Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Mouse anti-Human, BB515, Clone: 3A2-2E7, BD

Catalog No. BDB565225 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565225 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565225 Supplier BD Biosciences Supplier No. 565225
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunoglobulin (Ig) antigen receptors are composed of a non-covalently-associated complex of Ig and two other proteins, Ig α and Ig β , clustered as CD79a and CD79b, respectively. CD79b is a membrane glycoprotein of 229 residues, with a predicted relative molecular mass of 36-40 kDa. Its expression is restricted to B lineage cells, has been reported to associate with surface IgM receptor, and may be involved in signal transduction.

Antibody 3A2-2E7 (also known as SN8) was generated by immunization of cell membrane preparations from B-prolymphocytic leukemia cells. It has similar reactivity characteristics as clone CB3-1 (another CD79b available from BD Biosciences Pharmingen, Cat. No. 555678). Antibodies CD3-1 and 3A2-2E7 reacts with an epitope that is enhanced on certain B-cell leukemias such as prolymphocytic leukemia and lymphoma, but not on chronic lymphocytic leukemia.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79B
Applications Flow Cytometry
Classification Monoclonal
Clone 3A2-2E7
Conjugate Brilliant Blue 515 (BB515)
Description Igβ/SN8/3A2-2E7-1F5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cell membranes from human B-prol ymphocytic leukemia (B-PLL) cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.