Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8.1, Mouse, Clone: CD8-E1, Abnova™

Catalog No. 89134945 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-945 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-945 Supplier Abnova Corporation Supplier No. MAB1360
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD8.1.

Specifications

Antigen CD8.1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CD8-E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD8.1.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Mouse
Immunogen Native purified CD8.1 from mouse LN cells.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity React with mouse CD8.1.
Content And Storage Store lyophilized powder at -4°C. After reconstitution store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.